Human Recombinant MAPKAPK5 (GST Tag), Unactive (from Baculovirus (Insect cells))
Supplier: Sino Biological
M42-14G-100 M42-14G-1000 M42-14G-20
77559-984EA 1292.44 CAD
77559-984 77559-986 77559-988
Human Recombinant MAPKAPK5 (GST Tag), Unactive (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant full length Human MAPKAPK5 was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
  • Shipping temperature: Dry ice