Human Recombinant MAPKAPK2 (His Tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
M40-11H-50 M40-11H-500
77559-950EA 1845.44 CAD
77559-950 77559-952
Human Recombinant MAPKAPK2 (His Tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant Human MAPKAPK2 (46 to end) was expressed by Baculovirus in Sf9 insect cells using an N to terminal His tag.
  • Shipping temperature: Dry ice