Human Recombinant METTL11A, (GST Tag) (from Baculovirus (Insect cells))

Supplier: Sino Biological

M331-30G-100 M331-30G-1000 M331-30G-20
77559-762EA 1292.44 CAD
77559-762 77559-764 77559-766
Human Recombinant METTL11A, (GST Tag) (from Baculovirus (Insect cells))
Proteins and Peptides

Recombinant full length Human METTL11A was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.


  • Shipping temperature: Dry ice

New Product

Order Now

SPECIFICATIONS

Protein/peptide name NTMT1
Environmentally Preferable
Protein synonyms HOMT1A|NRMT|NTM1A|AD-003|METTL11A|C9orf32
Protein/peptide type Recombinant
Species Human
Source Baculovirus (Insect cells)
Conjugation Unconjugated
Formulation Supplied as sterile 50 mM Tris-HCl, pH 7.5, 50 - 300 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 - 1.0 mM DTT, 0 - 0.1 mM PMSF, 10 - 25% glycerol
Molecular weight ~51 kDa
Storage conditions Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
Tested applications Methyltransferase Assay

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR