Human Recombinant LZK (GST Tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
M21-11G-10 M21-11G-50 M21-11G-500
77559-582EA 549.84 CAD
77559-582 77559-584 77559-586
Human Recombinant LZK (GST Tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant Human LZK (1 to 427) was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
  • Shipping temperature: Dry ice