Human Recombinant MEF2A, (GST Tag) (from Baculovirus (Insect cells))
Supplier: Sino Biological
M20-34G-100 M20-34G-1000 M20-34G-20
77559-570EA 1624.24 CAD
77559-570 77559-572 77559-574
Human Recombinant MEF2A, (GST Tag) (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant full length Human MEF2A was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
  • Shipping temperature: Dry ice