Human Recombinant MLK3 (GST Tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
M19-11G-50 M19-11G-500
77559-560EA 1845.44 CAD
77559-560 77559-562
Human Recombinant MLK3 (GST Tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant Human MLK3 (1 to 488) was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
  • Shipping temperature: Dry ice