Human Recombinant COT (GST Tag), Unactive (from Baculovirus (Insect cells))
Supplier: Sino Biological
M16-15G-100 M16-15G-1000
77559-540EA 1292.44 CAD
77559-540 77559-542
Human Recombinant COT (GST Tag), Unactive (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant Human COT (30 to 397) was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
  • Shipping temperature: Dry ice