Human Recombinant TAK1-TAB1 (GST Tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
M15-13G-50 M15-13G-500
77559-530EA 1845.44 CAD
77559-530 77559-532
Human Recombinant TAK1-TAB1 (GST Tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant Human proteins TAK1 (1 to 303) and TAB1 (437 to end), linked by a small peptide (DFGGGGG), were expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
  • Shipping temperature: Dry ice