Human Recombinant MEK4 (MAP2K4, His Tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
M05-10H-50 M05-10H-500
77559-438EA 1845.44 CAD
77559-438 77559-440
Human Recombinant MEK4 (MAP2K4, His Tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant Human MKK4 (33 to end) was expressed by Baculovirus in Sf9 insect cells using an N to terminal His tag.
  • Shipping temperature: Dry ice