Human Recombinant LKB1(STK11, GST Tag) (from Baculovirus (Insect cells))
Supplier: Sino Biological
L15-30G-100 L15-30G-1000
77559-274EA 1292.44 CAD
77559-274 77559-276
Human Recombinant LKB1(STK11, GST Tag) (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant full length Human LKB1(STK11) was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
  • Shipping temperature: Dry ice