Human Recombinant LRRK1 (GST Tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
L09-11G-500 L09-11G-10 L09-11G-50
77559-188EA 4405.04 CAD
77559-188 77559-184 77559-186
Human Recombinant LRRK1 (GST Tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant Human LRRK1 (622 to 1610) was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
  • Shipping temperature: Dry ice