Human Recombinant KDM5C, (GST Tag) (from Baculovirus (Insect cells))
Supplier: Sino Biological
K425-31CG-100 K425-31CG-1000
77559-106EA 1292.44 CAD
77559-106 77559-108
Human Recombinant KDM5C, (GST Tag) (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant Human KDM5C (1 to 671) was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
  • Shipping temperature: Dry ice