Mouse Recombinant KDM5B, (GST Tag) (from Baculovirus (Insect cells))
Supplier: Sino Biological
K425-31BG-100 K425-31BG-1000
77559-100EA 1292.44 CAD
77559-100 77559-102
Mouse Recombinant KDM5B, (GST Tag) (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant mouse KDM5B (1 to 753) was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
  • Shipping temperature: Dry ice