Mouse Recombinant KDM4B, (GST Tag) (from Baculovirus (Insect cells))
Supplier: Sino Biological
K424-31BG-100 K424-31BG-1000
77559-082EA 1292.44 CAD
77559-082 77559-084
Mouse Recombinant KDM4B, (GST Tag) (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant mouse KDM4B (1 to 725) was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
  • Shipping temperature: Dry ice