Human Recombinant UHMK1 (KIS, GST Tag) (from Baculovirus (Insect cells))
Supplier: Sino Biological
K05-30G-100 K05-30G-1000
77558-740EA 1292.44 CAD
77558-740 77558-742
Human Recombinant UHMK1 (KIS, GST Tag) (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant full length Human UHMK1 (KIS) was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
  • Shipping temperature: Dry ice