Human Recombinant c-JUN, (GST Tag) (from E. coli)
Supplier: Sino Biological
J05-55G-100 J05-55G-1000
77558-670EA 1624.24 CAD
77558-670 77558-672
Human Recombinant c-JUN, (GST Tag) (from E. coli)
Proteins and Peptides
Recombinant Human c to JUN (1 to 79) was expressed in E. coli cells using a N to terminal GST tag.
  • Shipping temperature: Dry ice