Human Recombinant IkBA, (Full length, GST Tag) (from E. coli)
Supplier: Sino Biological
I20-30G-100 I20-30G-1000
77558-528EA 1624.24 CAD
77558-528 77558-530
Human Recombinant IkBA, (Full length, GST Tag) (from E. coli)
Proteins and Peptides
Recombinant full length Human IkBA was expressed in E. coli cells using an N to terminal GST tag.
  • Shipping temperature: Dry ice