QuEChERS Dispersive Solid Phase Extraction (SPE) Kits
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
5982-6662
5982-6663
5982-6664
5982-6665
AG5982-6662EA
388.68
CAD
AG5982-6662
AG5982-6663
AG5982-6664
AG5982-6665
QuEChERS Dispersive Solid Phase Extraction (SPE) Kits
Chromatography Sorbents
SPE Sorbents
QuEChERS dispersive solid phase extraction (SPE) kits supplied in 15 ml centrifuge tubes accommodate the aliquot volumes specified by current AOAC and EN methodologies.
- Non hazardous
The sorbent extracts the sample matrix, while MgSO₄ helps remove excess water and improve analyte partitioning.