Hei-PLATE Mix 'n' Heat Ultimate e-Lift Magnetic Hotplate Stirrers
Supplier: Heidolph NA, LLC
036110283 036110284
CA77506-888EA 4758.62 CAD
CA77506-888 CA77506-890
Hei-PLATE Mix 'n' Heat Ultimate e-Lift Magnetic Hotplate Stirrers
Stirrers Magnetic Hotplate Stirrers
The Hei-PLATE Mix 'n' Heat Ultimate magnetic hotplate stirrers are equipped with numerous interfaces for automated laboratory processes and integration with the Hei-PROCESS software solution.
  • Package includes: Hei-PLATE Mix'n'Heat Ultimate magnetic hotplate stirrer for fast and precise heating without overshoot
  • Intuitive handling for digitalization
  • RS232 and USB B interfaces allow control via the Hei-PROCESS solution
  • LAN interface can directly integrate the device into a network
  • USB A interface located at the front allows data logging via USB stick
  • USB C interface compatible with Heidolph electronic automated lab jack, E-Lift
  • One display per main feature (stirring and heating)
  • Separate timers for heating and stirring functions
  • Three heating modes – FAST for quick heating and PRECISE and PRECISE+ for exact control of your target temperature without overshoot
  • Compatible with PT1000 and double sensor PT1000 for measurement in your sample and in the bath liquid simultaneously
  • 10 year-warranty
  • E-lift electrical lab jack for comfortable and secure work with a 3-year warranty
Hei-PLATE Mix 'n' Heat Ultimate is equipped with numerous interfaces for automated laboratory processes. The RS232 and USB B ports enable control via the integrated Hei-PROCESS solution. You can actually integrate your device in the Hei-CONTROL Pro software via the LAN interface. As from the middle of 2024, the LAN interface will additionally support the communication protocol OPC UA. The USB A interface at the front allows data logging via a USB stick. For more comfort and safety, you can connect the magnetic stirrer with the electric Heidolph lab jack via the USB C interface.
Certifications: TUV, NRTL, CSA and CE marked
Mechanics IP02, electronics IP42