GridION™ DNA/RNA Sequencing Devices
Supplier: Oxford Nanopore Technologies
GRD-MK1CAPX SLW12M-G SLW24M-G SLW36M-G SUPP001 GRDPROJECT96
77532-286EA 92825 CAD
77532-286 77532-294 77532-296 77532-298 77532-300 GRDPROJECT96
GridION™ DNA/RNA Sequencing Devices
Next Generation Sequencing Instruments
GridION ― a self-contained, easily deployable DNA/RNA benchtop nanopore sequencer.
  • Flexible — run up to five independently addressable flow cells
  • Integrated compute — powerful onboard data processing and analysis, minimizing IT requirements
  • Compact — small footprint, suitable for any lab
  • Real-time data analysis — rapid results with base modification data as standard
  • Any read length — from short to ultra-long (longest >4 Mb)
A flexible, self-contained, benchtop nanopore sequencer, running up to five MinION or Flongle Flow Cells (or combinations of each) that can respond to the needs of multiple users on demand, across varied applications. Integrated, high-performance data processing alleviates the need for complex IT infrastructure.
Ordering information: Choose a GridION™ purchase plan to suit your needs:
GridION™ Project Pack 96 enables users to get started with GridION™ with a consumable budget. The pack includes sequencing kits, flow cells, wash kits and control expansion kit (see ordering table for more information). A remote installation support call (Assurance) is provided as standard to ensure correct installation. GridION™ Project Pack components must be quoted separately.
GridION™ CapEx enables users to get started with GridION™ with a capital budget and therefore gain title to the device. A remote installation support call (Assurance) is provided as standard to ensure correct installation.
Note: A GridION™ Software License and Warranty must be purchased alongside each GridION™ device purchase.
GridION™ Advanced Training is also available to ensure the successful optimization of your nanopore sequencing experiments. This 2.5-day onsite training course for up to four participants will cover the essentials of nanopore sequencing — from planning your experiment through to sequencing and an introduction to data analysis. All training will be customised to your specific requirements and consumables (six flow cells and two sequencing kits) will be provided. Contact us to discuss your training needs.
For more information on nanopore sequencing library prep kits and flow cells, please visit the individual product pages.
Delivery information: Flow cells and sequencing kits provided with the GridION Project Pack 96 can be scheduled across three deliveries. Last delivery must be within nine months of device shipment. Contact us to discuss your package configuration requirements.
Caution: Not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.
Order Now
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.