Vacuum Pumps, V-80
Supplier: Buchi
11V080000 11V080001 11V080010 11V080011 11V080100 11V080101 11V080110 11V080111 11V080200 11V080201 11V080210 11V080211
77569-888EA 4577.17 CAD
77569-888 77569-890 77569-892 77569-894 77569-896 77569-898 77569-900 77569-902 77569-904 77569-906 77569-908 77569-910
Vacuum Pumps, V-80
Pumps Vacuum Pumps
The vacuum pump V-80 is a chemically resistant PTFE diaphragm vacuum pump with space saving design. It covers essential needs in vacuum generation.
  • Effective vacuum source for limited budgets
  • Small footprint saves valuable laboratory space
  • Light weight and compact design with removable carrying handle
  • High up time due to low maintenance
  • Accurate vacuum regulation thanks to optional interface
  • Robust and stable design and Integrated ECO2 mode
  • On/off-regulated by the Interface I-180, optimally suited to work with Rotavapor® R-80
Speed-controlled vacuum solutions: The optional Vacuum Pump V-80/V-180 features precise speed control, ensuring a quiet vacuum generation and reduced bumping. The Vacuum Interface I-80/I-180 provides an easy means for vacuum control and can conveniently be attached directly to the Rotavapor®®, the pump, or a lab stand for optimal flexibility.
Certifications: CE, CSA Certified.
Delivery information: Includes silencer and 2 m of vacuum tubing.