Water Ultra Pure, CellPro™, Molecular Biology Grade Ultra Pure

Supplier: Alkali Scientific

DC42111 DC42311-4L
104026-888EA 546.15 CAD
104026-888 104027-060
Water Ultra Pure, CellPro™, Molecular Biology Grade Ultra Pure
Water
Formula: H₂O
MDL Number: MFCD00011332
CAS Number: 7732-18-5
UN: 3248

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR