Supreme Frosted Plus Microscope Slides
Supplier: Alkali Scientific
SM2575-G SM2575-O SM2575-Y SM2576 SM2576-O SM2575-B
104026-768EA 641.54 CAD
104026-768 104026-946 104028-680 104026-988 104027-898 104028-058
Supreme Frosted Plus Microscope Slides
Slides Microscope Slides
Enhance visibility and sample adherence with Supreme Frosted Plus microscope slides, ideal for a variety of histology and cytology applications.
  • Frosted
  • Positive electrical charge
  • Glass
  • Cryosectioning
  • Whole mount staining
Supreme Frosted Plus microscope slides are meticulously designed to meet the demanding requirements of laboratory applications, offering exceptional quality and performance. Each slide features frosted edges and a positive electrical charge, ensuring secure sample adhesion and reliable results. Crafted from crystal-clear glass, theseMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable