Radiant™ Probe 1-Step Lo-ROX qPCR Kits
Supplier: Alkali Scientific
QL4001 QL4003 QL4012
104028-446EA 169.23 CAD
104028-446 104027-982 104027-428
Radiant™ Probe 1-Step Lo-ROX qPCR Kits
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Perform quantitative PCR assays with Radiant™ Probe 1-Step Lo-ROX qPCR Kits, ensuring reliable and sensitive detection.
  • Novel hot-start chemistry for improved specificity and sensitivity
  • Next-generation PCR buffer formulations for maximum PCR efficiency and reaction speed
  • Broad range detection for increased reliability in low-copy assays
Radiant™ Probe 1-Step Lo-ROX qPCR Kits, a high-performance portfolio of 2X ready-to-use real-time PCR reagents designed for best-in- class quantitative PCR. Radiant™ qPCR Kits are engineered for robust real-time PCR with earlier quantification cycle values (Ct), industry-leading sensitivity (increased limit of detection) and exceptional speed (rapid extension rates). The buffer system allows highly efficient amplification of GC-rich and AT-rich sequences in addition to dramatic improvements in PCR sensitivity in low-copy assays and PCR conditions conducive to ultra-fast amplification. The Radiant™ Probe 1-Step Lo-ROX qPCR Kit utilizes a proprietary, Green non-inhibitory intercalating dye for robust, fluorescent detection.