Radiant™ Probe 1-Step Hi-ROX qPCR Kits
Supplier: Alkali Scientific
QH5001 QH5003 QH5012
104027-960EA 232.31 CAD
104027-960 104026-966 104028-768
Radiant™ Probe 1-Step Hi-ROX qPCR Kits
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Utilize Radiant™ Probe 1-Step Hi-ROX qPCR Kits for precise quantitative PCR assays with probe-based detection.
  • Novel hot-start chemistry for improved specificity and sensitivity
  • Next-generation PCR buffer formulations for maximum PCR efficiency and reaction speed
  • Broad range detection for increased reliability in low-copy assays
Radiant™ Probe 1-Step Hi-ROX qPCR Kits, a high-performance portfolio of 2X ready-to-use real-time PCR reagents designed for best-in- class quantitative PCR. Radiant™ qPCR Kits are engineered for robust real-time PCR with earlier quantification cycle values (Ct), industry-leading sensitivity (increased limit of detection) and exceptional speed (rapid extension rates). The buffer system allows highly efficient amplification of GC-rich and AT-rich sequences in addition to dramatic improvements in PCR sensitivity in low-copy assays and PCR conditions conducive to ultra-fast amplification. The Radiant™ Probe 1-Step Hi-ROX qPCR Kit utilizes a proprietary, Green non-inhibitory intercalating dye for robust, fluorescent detection.