CellPro™ Thioglycolate, Molecular Biology Grade, Sterile Filtered
Supplier: Alkali Scientific
DC0009 DC0025
104028-784EA 46.15 CAD
104028-784 104027-402
CellPro™ Thioglycolate, Molecular Biology Grade, Sterile Filtered
Media for Microbiology Prepared Media for Microbiology
Promote anaerobic growth conditions with CellPro™ Thioglycolate, ensuring molecular biology grade purity and sterile filtration.
  • Recommended for the cultivation of aerobic, microaerophilic, and anaerobic microorganisms in normally sterile materials
CellPro™ Thioglycolate is a specialized medium designed for the cultivation of anaerobic and microaerophilic bacteria. With its unique formulation containing thioglycolate, dextrose, and other nutrients, this medium promotes bacterial growth under low-oxygen conditions. Each batch undergoes rigorous sterile filtration to maintain purity and consistency, providing researchers with a reliable tool for microbiological studies and microbial diversity analysis.