CellPro™ TAE Tris Acetate EDTA, Molecular Biology Grade, Sterile Filtered
Supplier: Alkali Scientific
DB0331 DB0331-CS DB0332-2L DB0332-CS
104027-322EA 67.69 CAD
104027-322 104027-892 104027-444 104026-858
CellPro™ TAE Tris Acetate EDTA, Molecular Biology Grade, Sterile Filtered
Buffers Electrophoresis Buffers Running Buffers
Prepare electrophoresis buffers with CellPro™ TAE Tris acetate EDTA, ensuring molecular biology grade purity and sterile filtration.
  • For molecular biology
  • 0.2 μm filtered
  • Sterile
  • Ultra pure
CellPro™ TAE Tris acetate EDTA is a fundamental reagent in nucleic acid electrophoresis, renowned for its exceptional buffering capacity and DNA solubilization properties. Designed to provide optimal performance and reproducibility, this solution enables precise separation and analysis of DNA fragments in agarose gel electrophoresis. Each batch undergoes rigorous sterile filtration to ensure purity and consistency, empowering researchers with confidence in their molecular biology experiments.