CellPro™ Tris Glycine, Sterile
Supplier: Alkali Scientific
DB0256 DB0241 DB0347-CS DB0348 DB0348-CS DB0349 PB0301
104027-652EA 43.08 CAD
104027-652 104027-068 104028-804 104027-904 104028-196 104027-142 104027-226
CellPro™ Tris Glycine, Sterile
Buffers Electrophoresis Buffers Running Buffers
Optimize protein transfer with CellPro™ Tris glycine buffer, ensuring reliability and sterility in your experiments.
  • For preparation of protein samples for native gel electrophoresis
  • Ultra pure
CellPro™ Tris glycine offers a sterile solution ideal for a variety of biochemical and molecular biology applications. Composed of Tris (Tris(hydroxymethyl)aminomethane) and Glycine, this buffer provides optimal conditions for protein electrophoresis, particularly in SDS-PAGE (Sodium dodecyl sulfate polyacrylamide gel electrophoresis) systems. The sterile formulation ensures the purity and integrity of your samples, minimizing the risk of contamination and ensuring reliable results. CellPro™ Tris glycine buffer is versatile and suitable for use in protein separation, transfer, and analysis techniques, making it an essential component in the laboratory for protein research and characterization.