Ultra Low Attachment Plates, Individually Wrapped, Sterile
Supplier: Alkali Scientific
CFP06 CFP24 CFP96
104026-738EA 146.15 CAD
104026-738 104026-996 104027-196
Ultra Low Attachment Plates, Individually Wrapped, Sterile
Microplates Cell Culture Plates
Maintain sterile conditions with individually wrapped, sterile ultra low attachment plates, ideal for sensitive cell culture applications.
  • Ultra low cell adhesion
  • Comprised of a covalently bound hydrogel layer that minimizes cell attachment, protein absorption and cellular activation
  • Use when culturing stem cells and regenerative cell therapy
  • Promotes growth of suspension cell lines - both established and primary cells
  • Ideal environment to help nurture embryoid body formation from ES cells
  • Product Shelf Life: 3 years at room temperature storage (25 °C)
Ultra low attachment plates provides an optimized environment for 3D cell culture. These high quality culture vessels are extremely effective for the culturing of spheroid cells. They do not require any special incubation techniques, and thus 3D cell culture can easily be implemented in the same way as conventional 2D culture. Ultra-low attachment plastics are manufactured from optically clear virgin polystyrene and are supplied with lot numbers traceability under the most demanding and sensitive cell culture work. Comprised of covalently bound hydrogel layer that minimizes cell attachment, protein absorption and cellular activation. The ultra low attachment plates and dishes are ideal for culturing stem cells and use during regenerative cell therapy. Promotes growth of suspension cell lines - both established and primary cells. Ideal environment to help nurture embryoid body formation from ES cells.