CellPro™ Gel Loading Dye
Supplier: Alkali Scientific
AG5002 AG6000
104028-122EA 180 CAD
104028-122 104028-790
CellPro™ Gel Loading Dye
Stains Electrophoresis Stains
Simplify gel loading procedures with CellPro™ gel loading dye.
  • No DNA masking during gel exposure to UV light
  • EDTA binds divalent metal ions and inhibits metal dependent nucleases
CellPro™ Gel loading dye is a specialized solution used in gel electrophoresis procedures for visualizing DNA or RNA samples during analysis. This loading dye contains tracking dyes to aid in monitoring the progress of electrophoresis and ensuring accurate loading of samples into the gel wells. Its formulation helps to enhance sample visibility and migration, facilitating precise analysis and interpretation of results.