Benchmark Scientific™ ST45 SureTherm CO₂ Incubators
Supplier: Alkali Scientific
RS8090 RS8091
104027-614EA 11784.62 CAD
104027-614 104027-272
Benchmark Scientific™ ST45 SureTherm CO₂ Incubators
Incubators CO₂ Incubators
Ensure precise CO₂ regulation with Benchmark Scientific™ ST45 SureTherm CO₂ Incubators, equipped with two shelves for optimal sample storage.
  • Precision temperature control, 6-sided heating
  • Precise monitoring over CO₂ with dual beam IR sensor
  • Seamless, stainless-steel chamber
  • Internal Microscope option
  • Excellent accuracy and recovery
  • Heated door and door frame
  • Digital control of humidity percentage (PLUS model only)
With excellent heat distribution technology, the ST Series of CO₂ incubators provide the highest levels of temperature accuracy with minimal fluctuation (within 0.1 °C). The six-side heating system, in combination with a low speed internal fan, uniformly spreads air throughout the chamber (without air exchange from the external environment). The chamber walls are constructed of high grade (316) stainless-steel, and all corners are rounded, preventing the accumulation of dirt/debris. For added security, all models feature an internal alarm to alert the user if the CO₂ percentage changes, or the temperature fluctuates by more than 1 °C from the set point. In addition, a 'quick-cal' calibration button allows quickly adjusting the chamber temperature to match that of a properly calibrated external thermometer. Two series (standard and PLUS) are each available in 45 and 180 L chamber sizes.
The PLUS model includes advanced programming for high heat decontamination (at 125 °C), a split inner glass door provides access to the chamber without exposing it fully to the external environment and programmable humidity percentage, up to 95%.