QuickSilver™ Buffer Powder
Supplier: Alkali Scientific
RB1201 RB1202
104028-406EA 264.62 CAD
104028-406 104027-948
QuickSilver™ Buffer Powder
Buffers Electrophoresis Buffers Running Buffers
Prepare your buffers easily with QuickSilver™ buffer powder, ensuring high-quality solutions for your laboratory experiments.
  • Quick and economical
  • Convenient premeasured pouches
  • Each pouch makes 1 L of 1X buffer
  • No pH adjustment needed
  • Nuclease and protease free
  • Quality and consistency guaranteed
Nucleic Acid Electrophoresis Buffer
QuickSilver™ TAE (Tris-acetate-EDTA)
TAE is ideal for separation of large DNA fragments in agarose gels and is used as running buffer and in gel preparation. TAE is the buffer of choice when recovering DNA from gels and when downstream processing involves enzymatic reactions. Final concentration of a 1X working solution is 40 mM Tris, 20 mM acetic acid, 1 mM EDTA.
EB1000 QuickSilver™ TAE Buffer, 1X, 100 packs .
Protein Electrophoresis Buffers .
QuickSilver™ TGS (Tris-Glycine-SDS)
Tris-Glycine-SDS is one of the most common running buffers used for SDS-PAGE. A 1X solution contains 25 mM Tris, 192 mM Glycine and 0.1% SDS.
EB1100 QuickSilver™ Tris Glycine-SDS Buffer, 1X, 50 packs.
QuickSilver™ Tris-MES-SDS.
Small to medium molecular weight proteins can be resolved on SDS-PAGE gels using Tris-MES-SDS as a running buffer. Proteins resolve faster in gels using Tris-MES-SDS as a running buffer than those using Tris-MOPS-SDS. A 1X solution has a final concentration of 50 mM Tris, 50 mM MES, 0.1% SDS and 1 mM EDTA.
EB1101 QuickSilver™ Tris-MES-SDS Buffer, 1X, 50 packs.
QuickSilver™ Tris-MOPS-SDS
Tris-MOPS-SDS is ideal for use as a running buffer when resolving medium to large proteins with SDS-PAGE. Gels run with Tris-MOPS-SDS buffer run slower than those using Tris-MES-SDS buffer. The final concentration of a 1X working solution is 50 mM Tris, 50 mM MOPS, 0.1% SDS and 1 mM EDTA.
EB1102 QuickSilver™ Tris-MOPS-SDS, 1X, 50 packs.
Western Blotting, ELISA, Cell Culture, Chromatography
QuickSilver™ PBS and PBST Because its osmolality and ion concentration closely mimic that of the human body, PBS is commonly used for cell culture applications, as well as HPLC and MS. PBST is a wash buffer and diluent for ELISA. The 1X PBS solution contains 137 mN NaCl, 2.7 mM KCl and 10 mM phosphate. PBST is supplemented with 0.05% Tween-20.
EB1200 QuickSilver™ PBS Buffer, 1X, 100 packs.
EB1201 QuickSilver™ PBST Buffer, 1X, 100 packs.
QuickSilver™ TBS and TBST
TBS and TBST are used to maintain pH within a very narrow range for a variety of applications including western blotting and immunoassays. The final concentration of a 1X working solution is 137 mM NaCl, 2.7 mM KCl, 50 mM Tris. In addition, 1X TBST contains 0.1% Tween-20.
EB1202 QuickSilver™ TBS buffer, 1X, 100 packs.
EB1203 QuickSilver™ TBST buffer, 1X, 100 packs.