Radiant™ Probe qPCR Kit, Hi-ROX
Supplier: Alkali Scientific
QP8001 QP8005 QP8020 QP8100
104028-174EA 95.38 CAD
104028-174 104026-586 104029-054 104028-176
Radiant™ Probe qPCR Kit, Hi-ROX
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Achieve precise quantitative PCR results with Radiant™ Probe qPCR kit, Hi-ROX, optimized for probe-based detection.
  • Novel hot-start chemistry for improved specificity and sensitivity
  • Next-generation PCR buffer formulations for maximum PCR efficiency and reaction speed
  • Broad range detection for increased reliability in low-copy assays
  • Versatile 2X mixes, suitable for both ultra-fast and standard cycling methods
Radiant™ Probe Hi-ROX qPCR Kits, a high-performance portfolio of 2X ready-to-use real-time PCR reagents designed for quantitative PCR. Radiant™ qPCR Kits are engineered for robust real-time PCR with earlier quantification cycle values (Ct), industry-leading sensitivity (increased limit of detection) and exceptional speed (rapid extension rates). The buffer system allows highly efficient amplification of GC-rich and AT-rich sequences in addition to dramatic improvements in PCR sensitivity in low-copy assays and PCR conditions conducive to ultra-fast amplification. Furthermore, Radiant™ Probe Hi-ROX qPCR Kits are optimized for advanced multiplexing capability, delivering the same high efficiency, early Ct, and reaction speed even in complex multiplex assays.
Instrument Compatibility: The Radiant™ Probe Hi-ROX qPCR kit has been optimized for use with all probe chemistries including TaqMan™, FRET™, Scorpions and molecular beacon probes on qPCR instruments listed below. Each of these instruments has the capacity to analyze qPCR data with the passive reference signal either on or off.