Radiant™ Probe qPCR Kit, No-ROX
Supplier: Alkali Scientific
QP7001 QP7005 QP7020 QP7100 QP7500
104027-412EA 95.38 CAD
104027-412 104027-010 104028-888 104026-618 104028-682
Radiant™ Probe qPCR Kit, No-ROX
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Perform quantitative PCR assays conveniently with Radiant™ Probe qPCR kit, No-ROX, designed for probe-based detection without ROX normalization.
  • Novel hot-start chemistry for improved specificity and sensitivity
  • Next-generation PCR buffer formulations for maximum PCR efficiency and reaction speed
  • Broad range detection for increased reliability in low-copy assays
  • Versatile 2X mixes, suitable for both ultra-fast and standard cycling methods
Radiant™ Probe No-ROX qPCR Kits, a high-performance portfolio of 2X ready-to-use real-time PCR reagents designed for quantitative PCR. Radiant™ qPCR Kits are engineered for robust real-time PCR with earlier quantification cycle values (Ct), industry-leading sensitivity (increased limit of detection) and exceptional speed (rapid extension rates). The buffer system allows highly efficient amplification of GC-rich and AT-rich sequences in addition to dramatic improvements in PCR sensitivity in low-copy assays and PCR conditions conducive to ultra-fast amplification. Furthermore, Radiant™ Probe No-ROX qPCR Kits are optimized for advanced multiplexing capability, delivering the same high efficiency, early Ct, and reaction speed even in complex multiplex assays.
Instrument Compatibility: The Radiant™ Probe No-ROX qPCR Kit has been optimized for use with all probe chemistries including TaqMan™, FRET™, Scorpions and molecular beacon probes on qPCR instruments that do not require the use of ROX for data normalization such as Bio-Rad iCycler®, iQTM5, MyIQ™, Opticon™, Opticon2™, MiniOpticon, Chromo4™, CFX96, CFX384, Roche LightCycler®480, Qiagen Rotor-Gene™ 3000 and 6000, Eppendorf Mastercycler®ep realplex, Cepheid SmartCycler™, and Illumina® Eco™.