ProCycle™ White Roche 480 LightCycler® qPCR Plate, Full Edge
Supplier: Alkali Scientific
PC9R PC3R
104027-804EA 135.38 CAD
104027-804 104028-638
ProCycle™ White Roche 480 LightCycler® qPCR Plate, Full Edge
Microplates PCR Plates
Conduct quantitative PCR experiments with precision using ProCycle™ White Roche 480 LightCycler® qPCR Plate, featuring a full edge design, available in packs of 10.
  • Broad thermal cycler compatibility
  • High-contrast, black alphanumeric indexing for easy well identification
  • Ultra-thin, uniform wells ensure optimal heat transfer for high reaction efficiency
  • Well ridges reduce sample evaporation when sealing with film, foil, or strip caps
  • Manufactured from high-clarity, ultra-pure polypropylene (meets FDA 21 CFR 177.1520 and USP Class VI)
  • For use in pharmaceutical industry, food industry, and molecular biology
  • Certified free of detectable DNase, RNase, DNA, PCR inhibitors, and tested pyrogen-free
Experience superior performance in quantitative PCR (qPCR) with ProCycle™ White Roche 480 LightCycler® qPCR Plates. These plates are specifically designed for use with the Roche 480 LightCycler® system, offering precise and reliable results in gene expression analysis, genotyping, and pathogen detection.
With a full-edge design and optimal thermal conductivity, these plates ensure uniform heating and efficient PCR amplification across all wells. Each pack contains 10 plates, providing ample supply for your high-throughput qPCR experiments. ProCycle™ qPCR Plates for accurate and reproducible results in your molecular diagnostics and research applications.