CellPro™ LB Media, Molecular Biology Grade, Sterile Filtered
Supplier: Alkali Scientific
DC0005 DC0026 DC0030 DC0015 DC0016 DC0003 DC0004
104028-036EA 83.08 CAD
104028-036 104028-082 104028-172 104027-056 104028-910 104028-562 104028-236
CellPro™ LB Media, Molecular Biology Grade, Sterile Filtered
Media for Microbiology Prepared Media for Microbiology
Cultivate robust bacterial cultures with CellPro™ LB media, meeting molecular biology grade standards and provided sterile filtered for purity.
  • Filter-sterilized, ready-to-use format
  • Convenient package size
  • Standard formulation
CellPro™ LB media is a rich bacterial growth medium widely used in microbiology and molecular biology research. With its balanced composition of peptone, yeast extract, and sodium chloride, this media supports robust bacterial growth and protein expression. Each batch undergoes rigorous sterile filtration to maintain purity and consistency, providing researchers with a reliable tool for bacterial culture, protein production, and recombinant DNA technology.