CellPro™ TE 1X (Tris EDTA), Molecular Biology Grade
Supplier: Alkali Scientific
DB0305 DB0306 DB0308 DB0263
104026-864EA 110.77 CAD
104026-864 104027-816 104026-658 104027-758
CellPro™ TE 1X (Tris EDTA), Molecular Biology Grade
Buffers Biological Buffers
Prepare reliable DNA and RNA solutions with CellPro™ TE 1X (Tris EDTA), meeting molecular biology grade standards for purity and reliability.
  • Molecular biology grade
  • RNase-free, economical, and ready-to-use
  • Sterile
  • Filtered
Introducing CellPro™ TE 1X (Tris EDTA), meticulously formulated by to meet the exacting standards of molecular biology research. This molecular biology grade TE buffer ensures exceptional purity and performance, making it an indispensable tool for various molecular biology applications, including DNA and RNA purification, DNA sequencing, and PCR.
With its precisely balanced composition and stringent quality control measures, CellPro™ TE 1X provides researchers with reliable and reproducible results, facilitating accurate and efficient laboratory workflows. CellPro™ TE 1X for molecular biology experiments and experience the assurance of superior quality and performance in every assay.