CellPro™ HEPES Buffer, Sterile, Filtered
Supplier: Alkali Scientific
Maintain pH stability in cell culture with CellPro™ HEPES buffer, providing sterile and filtered solutions for reliable results.
- High purity product for biochemical and biological research
- Suitable for cell culture
- High solubility membrane impermeability
- Limited effect on biochemical reactions
- Very low visible and ultraviolet light absorbance
- Chemically and enzymatically stable
- Easy to prepare
CellPro™ HEPES buffer is a vital component in laboratory protocols, offering a sterile and filtered solution indispensable for various biochemical and cell culture applications. Engineered with precision, this buffer solution ensures the purity and consistency required for reliable research outcomes. Its sterile nature minimizes the risk of contamination, making it suitable for cell culture maintenance, protein assays, and molecular biology experiments.
With a filtered formulation, CellPro™ HEPES buffer guarantees clarity and reliability in experimental results, providing researchers with confidence in their findings. Whether used for maintaining optimal pH levels in cell culture media or as a buffering agent in biochemical assays, this solution offers a dependable option for diverse laboratory needs. Trust CellPro™ HEPES Buffer to deliver consistent performance, supporting the integrity of scientific investigations and the pursuit of new discoveries.