Radiant™ DNA Markers and Ladders I to IV

Supplier: Alkali Scientific

C614 C616
104028-962EA 113.85 CAD
104028-962 104026-802
Radiant™ DNA Markers and Ladders I to IV
Electrophoresis Reagents Molecular Weight Markers

Achieve accurate sizing and analysis with Radiant™ DNA markers and ladders I to IV, trusted standards for molecular biology research.


  • Reliable PCR grade
  • Convenient, pre-optimized and pre-mixed
  • Ultra-pure: >99% trisphosphate by HPLC
  • Extended shelf-life of 18 months at −20 °C
  • Free from PCR inhibitors
  • DNase, RNase and Nickase free
  • Manufactured by ASI in a purpose-built facility
  • Applications: standard and long range PCR assays, cDNA synthesis, qPCR, microarrays, DNA sequencing, labeling


A ready-to-use molecular grade dNTP mix containing dATP, dCTP, dGTP and dTTP at pH 7.5 as sodium salts in purified water.


The DNA markers and ladders I to IV is designed to save hands-on time for researchers and minimize the possibility of contamination. For use in DNA polymerization reactions, DNA labeling and sequencing processes.


All ASI dNTPs are supplied as sodium salts in purified water at pH 7.5. Sodium salts have resistance to repeated freezing and thawing cycles. Also dNTP preparations remain sterile over the entire shelf-life due to the bacteriostatic activity of sodium towards various microorganisms. dNTPs Mol. Water Molar Extinction (1 µmol) dATP 514.9 15.4 (A259) dGTP 530.9 13.7 (A252) dCTP 490.9 9.1 (A272) dTTP 505.9 9.6 (A267).

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR