Lakeland CleanMax® Select Disposable Cleanroom Frocks, SMMS
Supplier: Lakeland
SBC191CS-2X SBC191CS-3X SBC191CS-4X SBC191CS-5X SBC191CS-LG SBC191CS-MD SBC191CS-SM SBC191CS-XL SBC191CM-2X SBC191CM-3X SBC191CM-4X SBC191CM-5X SBC191CMB-2X SBC191CMB-3X SBC191CMB-4X SBC191CMB-5X SBC191CMB-LG SBC191CMB-MD SBC191CMB-SM SBC191CMB-XL SBC191CM-LG SBC191CM-MD SBC191CM-SM SBC191CM-XL
77188-322CS 431.09 CAD
77188-322 77188-324 77188-326 77188-328 77188-330 77188-332 77188-334 77188-336 77188-290 77188-292 77188-294 77188-296 77188-298 77188-300 77188-302 77188-304 77188-306 77188-308 77188-310 77188-312 77188-314 77188-316 77188-318 77188-320
Lakeland CleanMax® Select Disposable Cleanroom Frocks, SMMS
Disposable Garments
The Lakeland CleanMax® Select frock is designed with breathable SMMS fabric that is ideal for ISO Class 5 to 8 cleanrooms.
  • Breathable SMMS fabric
  • Non-sterile and sterile options available
  • Tunnelized, latex-free elastic
  • Low linting
  • Elastic wrists
  • Thumb loops
CleanMax® Select SMMS is available in sterile and non-sterile configurations. Clean-manufactured in Lakeland owned and operated certified cleanrooms, under strict process and environmental controls CleanMax® Select meets IEST particle category cleanliness levels. CleanMax® Select SMMS sterileMore Product Information