CellPro™ Trypticase Soy Agar
Supplier: Alkali Scientific
DC0136 DC0137
104027-488EA 80 CAD
104027-488 104029-030
CellPro™ Trypticase Soy Agar
Media for Microbiology Prepared Media for Microbiology
Ensure reliable bacterial growth with CellPro™ Trypticase soy agar, providing a nutrient-rich environment for microbial culture.
  • General purpose, non-selective media for growing wide range of organisms
CellPro™ Trypticase Soy Agar is a general-purpose medium used for the cultivation and isolation of a wide range of microorganisms in clinical and laboratory settings. This agar medium is formulated with enzymatic digest of casein (tryptone), enzymatic digest of soybean meal (soytone), sodium chloride, agar, and other nutrients. Trypticase Soy Agar supports the growth of both fastidious and non-fastidious bacteria, making it suitable for various microbiological applications, including environmental monitoring, food testing, and pharmaceutical quality control. The plates (Ø 100 mm ) provide a convenient platform for bacterial culture experiments and microbial enumeration.