CellPro™ LB Agar, Miller
Supplier: Alkali Scientific
DC0141 DC0129 DC0130 DC0131 DC2620 DC2630 DC0097 DC0103 DC0105 DC0106 DC0044 DC0045 DC0109 DC0110 DC0112 DC0114 DC0155 DC0156 DC0157 DC0158 DC0160 DC0125 DC0126 DC0127 DC0162 DC0164 DC0166 DC0167 DC0093 DC0094
104026-960EA 116.92 CAD
104026-960 104028-764 104027-174 104027-564 104027-688 104028-836 104026-746 104027-590 104027-160 104028-228 104028-128 104029-006 104026-692 104028-866 104027-528 104027-672 104027-680 104028-338 104026-726 104028-160 104026-872 104028-960 104027-908 104027-732 104027-694 104028-010 104028-940 104026-978 104027-578 104027-632
CellPro™ LB Agar, Miller
Media for Microbiology Prepared Media for Microbiology
Prepare nutrient-rich media with CellPro™ LB Agar, Miller, providing essential components for bacterial growth and colony formation.
  • Elevate your bacterial culturing with CellPro™ premium LB Agar, Miller formulations
  • Ensure optimal growth and colony formation for your microbiology experiments
  • Experience reliable performance and consistent results for your research needs
CellPro™ LB Agar, Miller is a solid medium commonly used for the cultivation and maintenance of Escherichia coli (E. coli) and other recombinant bacterial strains. This agar variant contains tryptone, yeast extract, sodium chloride, and agar, providing essential nutrients for bacterial growth. LB Agar, Miller is particularly suitable for molecular biology and microbiology applications, including the selection and propagation of transformed bacterial cells carrying recombinant DNA constructs.