Cell Culture Chamber Slides
Supplier: Alkali Scientific
TCC001 TCC002 TCC004 TCC008
104028-490EA 384.62 CAD
104028-490 104027-616 104027-120 104028-488
Cell Culture Chamber Slides
Slides Chamber Slides
Create controlled cell culture environments with these cell culture chamber slides.
  • Alphanumeric well identification
  • Silicone gasket provides leak-free operation
  • Oversized hinged clasps provide easy handling and more uniform pressure
  • Pyrogen/cytotoxin-free
  • Gamma sterilized
These Cell culture chamber slides, available in a case of 12, provide convenient and reliable platforms for various cell culture applications. With individual chambers designed to accommodate cell growth and experimentation, these slides facilitate parallel processing and standardized assays, enhancing experimental throughput and efficiency.