Premium Scar Scratchers
Supplier: Alkali Scientific
SCAR25 SCAR26 SCAR6 SCAR24
104028-652EA 156.92 CAD
104028-652 104028-850 104026-778 104028-212
Premium Scar Scratchers
Spreaders Cell Spreaders
Facilitate cell research with Alkali Scientific™ Premium Scar Scratchers.
  • Non pyrogenic
  • Non cytotoxic
  • DNase/RNase free
  • Human DNA free
  • Sterile
The Scar Scratcher is a specialized tool designed to enhance the uniformity and reproducibility of outcomes in research related to cancer metastasis, wound healing and cell migration. Its innovative design and precise functionality make it an invaluable instrument for investigators aiming to study these critical biological processes with accuracy and consistency.
Packaging: 20 per case.