Radiant™ SYBR Green Master Mix qPCR Kit, Hi-ROX
Supplier: Alkali Scientific
Achieve robust quantitative PCR results with Alkali Scientific™ Radiant™ SYBR Green Master Mix qPCR Kit, Hi-ROX, optimized for sensitive detection with high ROX content.
- Next-generation PCR buffer formulations for maximum PCR efficiency and reaction speed
- Novel hot-start chemistry for improved specificity and sensitivity
- Broad range detection for increased reliability in low-copy assays
- Versatile 2X Mixes, suitable for both ultra-fast and standard cycling methods
Alkali Scientific offers Radiant™ Green Hi-ROX qPCR Kits, a high-performance 2X ready-to-use real-time PCR reagents designed for quantitative PCR. Radiant™ Green Hi-ROX qPCR Kits are engineered for robust real-time PCR with earlier quantification cycle values (Ct), industry-leading sensitivity (increased limit of detection) and exceptional speed (rapid extension rates). The buffer system allows highly efficient amplification of GC-rich and AT-rich sequences in addition to dramatic improvements in PCR sensitivity in low-copy assays and PCR conditions conducive to ultra-fast amplification. The Radiant™ Green Hi-ROX qPCR Kit utilizes a proprietary, Green non-inhibitory intercalating dye for robust, fluorescent detection in combination with fluorescein for optional use with certain qPCR instruments.
Instrument Compatibility:
The Radiant™ Green Hi-ROX qPCR Kit has been optimized for use with intercalating-dye based real-time PCR. Each of these instruments has the capacity to analyze qPCR data with the passive reference signal either on or off.