Radiant™ Probe qPCR Kit, Lo-ROX
Supplier: Alkali Scientific
QP9001 QP9005 QP9100
104028-712EA 95.38 CAD
104028-712 104027-534 104028-124
Radiant™ Probe qPCR Kit, Lo-ROX
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Perform sensitive quantitative PCR assays with Alkali Scientific™ Radiant™ Probe qPCR Kit, Lo-ROX, designed for reliable detection with low ROX content.
  • Novel hot-start chemistry for improved specificity and sensitivity
  • Next-generation PCR buffer formulations for maximum PCR efficiency and reaction speed
  • Broad range detection for increased reliability in low-copy assays
  • Versatile 2X mixes, suitable for both ultra-fast and standard cycling methods
Alkali Scientific offers Radiant™ Probe Lo-ROX qPCR Kits, a high-performance portfolio of 2X ready-to-use real-time PCR reagents designed for quantitative PCR. Radiant™ qPCR Kits are engineered for robust real-time PCR with earlier quantification cycle values (Ct), industry-leading sensitivity (increased limit of detection) and exceptional speed (rapid extension rates). The buffer system allows highly efficient amplification of GC-rich and AT-rich sequences in addition to dramatic improvements in PCR sensitivity in low-copy assays and PCR conditions conducive to ultra-fast amplification. Furthermore, Radiant™ Probe Lo-ROX qPCR Kits are optimized for advanced multiplexing capability, delivering the same high efficiency, early Ct, and reaction speed even in complex multiplex assays.
Instrument Compatibility:
The Radiant™ Probe Lo-ROX qPCR Kit has been optimized for use with all probe chemistries including TaqMan™, FRET™, Scorpions and molecular beacon probes on qPCR instruments listed below. Each of these instruments has the capacity to analyze qPCR data with the passive reference signal either on or off. The Kit is also compatible with several instruments that do not require the use of ROX, such as the Bio-Rad CFX96, Roche LightCycler® 480, or the Qiagen Rotor-Gene® 6000.