CellPro™ Premium Cell Strainer
Supplier: Alkali Scientific
MT4040 MT4070 MT4100
104027-522EA 181.54 CAD
104027-522 104028-238 104026-898
CellPro™ Premium Cell Strainer
Cell Strainers
Ensure efficient cell straining with CellPro™ Premium Cell Strainer, available in packs of 50.
  • Advanced design - faster flow through
  • Compatible with 6-well plates
  • Interlocking and stackable
  • Ga mma sterilized, individually wrapped
  • RNase-, DNase-, pyrogen-, and cytotoxin-free
  • Four 1.5 mm feet for straining into flat plates and dishes
  • Designed for use with 50 ml tubes
  • Adapter for 15 ml tubes included
Premium cell strainer system is a fast and easy-to-use solution for detaching, isolating, and straining cells from clusters and tissues. A polypropylene frame supports a nylon mesh that is available in three color-coded porosities: 40 μm (blue), 70 μm (white), and 100 μm (yellow). The advanced frame/housing design also contributes to superior performance and reduced processing time, by eliminating the 'air-lock' effect. Improved aeration allows air to escape while filtrate is entering the tube, preventing messy spillovers.