CIP Neutralizer® Alkaline Based Neutralizers
Supplier: Steris
1D07D3
77531-110EA 12303.46 CAD
77531-110
CIP Neutralizer® Alkaline Based Neutralizers
Cleaning Agents Detergents and Surface Cleansers
CIP Neutralizer® Alkaline neutralizer is a liquid blend of sodium hydroxide and potassium hydroxide suitable for a variety of applications where raising the pH or increasing the alkalinity of a solution is required.
  • Productivity - Raises the pH of solution utilizing a blend of potassium and sodium hydroxide
  • Environmentally friendly - Formulated with low mercury content in order to meet environmental requirements
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility
CIP Neutralizer® Alkaline based neutralizer is specially formulated for neutralization of acidic solutions.