CIP 130® Alkaline Process Cleaning Detergent

Supplier: Steris
CIP 130®
1C13D3 1C1305 1C1301
77531-114EA 5113.88 CAD
77531-114 77531-100 77531-096
CIP 130® Alkaline Process Cleaning Detergent
Cleaning Agents Detergents and Surface Cleansers
Process cleaning detergent for CIP, COP and manual applications.

  • Flexibility - Effective cleaning against a wide variety of pharmaceutical process residues in clean-in-place (CIP), clean-out-of-place (COP), and manual applications
  • Efficacy - Especially effective against oily and greasy residues due to multiple surfactant system
  • Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports
  • Cost savings - Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility

CIP 130® Alkaline Detergent is a proprietary blend of potassium hydroxide, an advanced surfactant system and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This low foaming product removes a wide range of process residues and is ideal for use in CIP, COP and manual applications.

CIP 130® Process Cleaning Detergent is specially formulated to meet the unique cleaning demands found in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.

CAS: 1310-58-3; 84133-50-6; 28805-58-5; 64-02-8
UN 1814
Hazard class: 8
Packing group: III

Caution: Danger, GHS05.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR