Mouse Recombinant 14-3-3 theta (His tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
Y84-30BH-100 Y84-30BH-1000 Y84-30BH-20
77565-592EA 1971.84 CAD
77565-592 77565-594 77565-596
Mouse Recombinant 14-3-3 theta (His tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length mouse 14-3-3 theta was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
  • Shipping temperature: Dry ice